missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYBB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CRYBB3 |
---|---|
Concentration | 0.49 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850595
|
Proteintech
21009-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840595
|
Proteintech
21009-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Crystallins are the domit structural components of the vertebrate eye lens.Specifications
CRYBB3 | |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P02524, P26998, Q9JJU9 | |
12962, 1417, 64349 | |
CRYBB3 Fusion Protein Ag15212 | |
Primary | |
-20°C | |
CRYBB3 |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Beta B3 crystallin, Beta crystallin B3, CATCN2, CRYB3, CRYBB3, crystallin, beta B3 | |
Crybb3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CRYBB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech