Learn More
Multiroir™ Crystal Polystyrene Minimax Storage Boxes
Use Multiroir™ Crystal Polystyrene Minimax Storage Boxes for your storage and transport needs. The clear, glass-like polystyrene enables you to view your contents clearly from the outside. The boxes are delivered complete with a separator kit for additional organization of contents.
Brand: Multiroir™ 42031
Additional Details : Weight : 0.24300kg
Description
- Transparent as glass
- Delivered complete with separator kit
- Various capacities
Specifications
Transparent | |
Rectangle | |
6 Case | |
228 mm |
Polystyrene | |
Storage Box | |
228 x 178 x 68 mm | |
68 mm |