Learn More
Invitrogen™ Human CSNK2A2 (CK2 Alpha 2), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

Specifications
Specifications
Accession Number | P19784 |
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 1459 |
Molecular Weight (g/mol) | 69.2 kDa |
Name | Human CSNK2A2 (CK2 Alpha 2), GST Tag |
Quantity | 100 μg |
Regulatory Status | RUO - research use only |
Gene Alias | 1110035J23Rik; C77789; casein kinase 2 alpha; casein kinase 2 alpha'; casein kinase 2 alpha 2; casein kinase 2, alpha prime polypeptide; casein kinase II subunit alpha'; casein kinase II, alpha 2, polypeptide; CK II alpha'; CK2; CK2A2; CK2alpha'; CSNK2A1; CSNK2A2 |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.