missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSRP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13432-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation.Specifications
CSRP1 | |
Polyclonal | |
Unconjugated | |
CSRP1 | |
CRP, CRP1, CSRP, CSRP1, CYRP, Cysteine rich protein 1, D1S181E | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13007, 1465, 29276 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21291, P47875, P97315 | |
Csrp1 | |
CSRP1 Fusion Protein Ag4236 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CSRP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction