missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTNNA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CTNNA3 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892424
|
Proteintech
13974-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882424
|
Proteintech
13974-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catenins are proteins that connect cadherin/beta-catenin cell adhesion complexes to the actin cytoskeleton. Although originally discovered in testis, alphaT-catenin is expressed in other tissues, the highest levels being observed in heart. Immunohistochemical analysis showed human alphaTcatenin localization at intercalated discs of cardiomyocytes and in peritubular myoid cells of testis. It is proven that alphaT-catenin can functionally restore cell-cell adhesion in colon cancer cells lacking alpha-catenins. This indicates that this protein is involved in the formation of specific cell-cell contacts in specific types of muscle cells. The encoded protein has about the same predicted size (100 kDa) as other alpha-catenins to which it shows an overall amino acid identity of 57%. In cells transfected with alphaT-catenin cDNA, interaction with alpha-catenin was demonstrated by coimmunoprecipitation. The 892_24D2S antibody against alphaT-catenin does not show any cross reaction with alphaE-catenin or alphaNcatenin in western blot analysis.Specifications
CTNNA3 | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q65CL1, Q9UI47 | |
216033, 29119, 361839 | |
CTNNA3 Fusion Protein Ag5008 | |
Primary | |
-20°C | |
CTNNA3 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Alpha T catenin, Cadherin associated protein, Catenin alpha 3, CTNNA3, VR22 | |
Ctnna3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CTNNA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech