missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTR9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20672-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognizes the C-term of CTR9.
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.Specifications
CTR9 | |
Polyclonal | |
Unconjugated | |
CTR9 | |
CTR9, KIAA0155, p150, p150TSP, SH2 domain binding protein 1, SH2BP1, TSBP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22083, 9646 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q62018, Q6PD62 | |
CTR9 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CTR9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction