missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTTNBP2NL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25523-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
CTTNBP2NL | |
Polyclonal | |
Unconjugated | |
CTTNBP2NL | |
CTTNBP2 N terminal like, CTTNBP2NL, KIAA1433 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55917, 80281 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99LJ0, Q9P2B4 | |
CTTNBP2NL | |
CTTNBP2NL Fusion Protein Ag22137 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |