missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTTNBP2NL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CTTNBP2NL |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824095
|
Proteintech
25523-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814095
|
Proteintech
25523-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The function of this protein remains unknown.Specifications
CTTNBP2NL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q99LJ0, Q9P2B4 | |
55917, 80281 | |
CTTNBP2NL Fusion Protein Ag22137 | |
Primary | |
-20°C | |
CTTNBP2NL |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CTTNBP2 N terminal like, CTTNBP2NL, KIAA1433 | |
CTTNBP2NL | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |