missing translation for 'onlineSavingsMsg'
Learn More

CTTNBP2NL Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25523-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824095

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The function of this protein remains unknown.
Specifications

Specifications

CTTNBP2NL
Polyclonal
Unconjugated
CTTNBP2NL
CTTNBP2 N terminal like, CTTNBP2NL, KIAA1433
Rabbit
Antigen Affinity Chromatography
RUO
55917, 80281
-20°C
Liquid
Western Blot, Immunoprecipitation
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99LJ0, Q9P2B4
CTTNBP2NL
CTTNBP2NL Fusion Protein Ag22137
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers