missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUGBP2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CUGBP2 |
---|---|
Concentration | 0.51 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890874
|
Proteintech
12921-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880874
|
Proteintech
12921-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CUG-BP1 and CUG-BP2 are proteins that bind specifically to (CUG)8 oligonucleotides in vitro. While CUG-BP1 has the major binding activity in normal cells, nuclear CUG-BP2 binding activity increases in DM cells. Both CUG-BP1 and CUG-BP2 are isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. CUG-BP1, an RNA CUG triplet repeat binding protein, regulates splicing and translation of various RNAs. Expansion of RNA CUG repeats in the DMPK in DM is associated with alterations in binding activity of CUG-BP1 as well as alterations in the translation of the C/EBPb transcription factor. CUG-BP1 is an important regulator of initiation from different AUG codons of C/EBPb mRNA.Specifications
CUGBP2 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Rat, Zebrafish, Mouse | |
O95319, Q6P0B1, Q792H5, Q9Z0H4 | |
10659, 14007, 29428, 373117 | |
CUGBP2 Fusion Protein Ag3594 | |
Primary | |
-20°C | |
CELF2 |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Bruno like protein 3, BRUNOL3, CELF 2, CELF2, CUG BP2, CUGBP2, ETR 3, ETR3, hNAPOR, NAPOR, RNA binding protein BRUNOL 3 | |
celf2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CUGBP2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech