Learn More
CUL1 Mouse anti-Human, Unconjugated, Clone: 1B6D10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66978-1-IG-150UL
 View more versions of this product
Product Code. 16879315
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The CUL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL1. This antibody recognizes human, mouse, rat antigen. The CUL1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CUL1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1B6D10
Unconjugated
150 μL
8454
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CUL1 Mouse anti-Human, Unconjugated, Clone: 1B6D10, Proteintech > 150 μL