Learn More
CUL1 Mouse anti-Human, Unconjugated, Clone: 1B6D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66978-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The CUL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL1. This antibody recognizes human, mouse, rat antigen. The CUL1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
CUL1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Mouse, Rat, Human |
Immunofluorescence, Immunohistochemistry, Western Blot | |
1B6D10 | |
Unconjugated | |
150 μL | |
8454 | |
IgG2b |