missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12895-1-AP-150UL
This item is not returnable.
View return policy
Description
CUL1 gene ontology annotations related to this gene include animal organ morphogenesis; apoptotic process; cell proliferation; protein monoubiquitination; SCF-dependent proteasomal ubiquitin-dependent protein catabolic process.Specifications
CUL1 | |
Polyclonal | |
Unconjugated | |
CUL1 | |
CUL 1, CUL1, cullin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8454 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13616 | |
CUL1 | |
CUL1 Fusion Protein Ag3585 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CUL1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction