Learn More
Custom PreAmp Pools
Custom PreAmp Pools
Brand: ABI Applied Biosystems 4459656
Additional Details : Weight : 0.00100kg
Description
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.Specifications
Custom TaqMan™ miRNA Pre-Amp Pool | |
Room Temperature | |
1 mL |
Format 16 | |
1 to 8 |