missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Custom PreAmp Pools
Custom PreAmp Pools
4070.00 SEK - 4220.00 SEK
Specifications
Type | Custom TaqMan™ miRNA Pre-Amp Pool |
---|---|
Content And Storage | Room Temperature |
No. of Samples | 1 to 8 |
Quantity | 1 mL |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
15504344
|
ABI Applied Biosystems
4459655 |
1 mL | Format 12 |
4215.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15514344
|
ABI Applied Biosystems
4459656 |
1 mL | Format 16 |
4220.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15524344
|
ABI Applied Biosystems
4459657 |
1 mL | Format 24 |
4140.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15534344
|
ABI Applied Biosystems
4459658 |
1 mL | Format 32 |
4210.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15544344
|
ABI Applied Biosystems
4459659 |
1 mL | Format 48 |
4140.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15554344
|
ABI Applied Biosystems
4459660 |
1 mL | Format 64 |
4210.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15564344
|
ABI Applied Biosystems
4459661 |
1 mL | 96A |
4220.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15574344
|
ABI Applied Biosystems
4459662 |
1 mL | 96B |
4070.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15504364
|
ABI Applied Biosystems
4478260 |
1 mL | Format 384 |
4215.00 SEK
1mL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
Description
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.Specifications
Custom TaqMan™ miRNA Pre-Amp Pool | |
Room Temperature | |
1 to 8 | |
1 mL |