Learn More
Custom PreAmp Pools
Custom PreAmp Pools
Brand:  ABI Applied Biosystems 4459660
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 15554344
  • 4210.00 SEK / 1mL
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.
Specifications
Specifications
Custom TaqMan™ miRNA Pre-Amp Pool
Room Temperature
1 mL
Format 64
1 to 8