Learn More
Invitrogen™ Custom Primer, 100% QC Illumina Each
Custom Primer, 100% QC Illumina Each
Brand:  Invitrogen™ 10668119
3.02 SEK valid until 2025-06-27
Use promo code "24112" to get your promotional price.
Product Code. 10543433
  • 4.53 SEK / Each
Nurture Your Progress
Use promo code 24108 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10ÎĽmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100% QC Illumina Primer, 25 nmol
100% QC Illumina Primer
Room Temperature
25 nmol
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Invitrogen™ Custom Primer, 100% QC Illumina Each >