Learn More
Invitrogen™ CX3CR1 Monoclonal Antibody (2A9-1), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 12-6099-42
Description
Description: This 2A9-1 monoclonal antibody reacts with human CX3CR1, which is the receptor for fractalkine, a transmembrane chemokine of the CX3C family. CX3CR1 is a seven transmembrane G protein-coupled receptor expressed on CD16+ NK cells, T cells (e.g. CD8+, CD4+, and gamma/delta), and monocytes. In non-immune cells, CX3CR1 has been found on osteoclast precursors and microglia. Little to no CX3CR1 surface expression can be detected on B cells and granulocytes. Together, fractalkine and its receptor mediate cell-cell adhesion and chemotaxis of NK cells, T cells, and monocytes. The expression of CX3CR1 has also been correlated with high levels of intracellular perforin and granzyme B. Applications Reported: This 2A9-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2A9-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 ÎĽL (0.125 ÎĽg) per test. A test is defined as the amount (ÎĽg) of antibody that will stain a cell sample in a final volume of 100 ÎĽL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 ÎĽm post-manufacturing filtered.
CX3CR1 is one of the chemokine receptors that are required as coreceptors for HIV infection. The genes encoding human, mouse, and rat CX3CR1 were cloned and designated V28 and CMKBRL1, CX3CR1, and RBS11, respectively. The encoded seven transmembrane protein was recently identified as the receptor for a novel transmembrane molecule, fractalkine, and renamed CX3CR1. Recently, CX3CR1 was found to serve as a coreceptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by fractokine. CX3CR1 mediates leukocyte migration an d adhesion. CX3CR1 is expressed in a variety of human tissues and cell lines.
Specifications
CX3CR1 | |
Monoclonal | |
5 ÎĽL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P49238 | |
CX3CR1 | |
Affinity chromatography | |
RUO | |
1524 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2A9-1 | |
PE | |
CX3CR1 | |
Beta chemokine receptor-like 1; CCRL1; chemokine (C-C) receptor-like 1; chemokine (C-X3-C motif) receptor 1; chemokine (C-X3-C) receptor 1; CMKBRL1; CMK-BRL1; CMK-BRL-1; CMKDR1; Cx3c; CX3C chemokine receptor 1; C-X3-C CKR-1; C-X3-C motif chemokine receptor 1; Cx3cr1; Fractalkine receptor; G protein-coupled receptor 13; GPR13; G-protein coupled receptor 13; GPRV28; RBS11; Scyd1; V28 | |
Rat | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.