missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5R1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11807-1-AP-150UL
This item is not returnable.
View return policy
Description
NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.Specifications
CYB5R1 | |
Polyclonal | |
Unconjugated | |
CYB5R1 | |
B5R.1, CYB5R1, cytochrome b5 reductase 1, humb5R2, NADH cytochrome b5 reductase 1, NQO3A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304805, 51706, 72017 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5EB81, Q9DB73, Q9UHQ9 | |
Cyb5r1 | |
CYB5R1 Fusion Protein Ag2475 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CYB5R1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction