Läs mer
Cyclo(L-prolyl-L-leucyl), TRC
Produktkod. 30354915
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354915 50 mg
30356785 10 mg
30368384 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98766050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2873-36-1
Molekylformel C11 H18 N2 O2
Molekylvikt (g/mol) 210.27
Synonym (3S-trans)-Hexahydro-3-(2-methylpropyl)-pyrrolo[1,2-a]pyrazine-1,4-dione, 3-Isobutylhexahydropyrrolo [1,2-a] pyrazine-1,4-dione, Cyclo(L-Leu-L-Pro), Cyclo(L-Pro-D-Leu), Cyclo(L-Pro-L-Leu), Cyclo(L-leucyl-L-prolyl), Cyclo(L-prolyl-L-leucyl), Cyclo(Pro-Leu), Cyclo(proline-leucine), Cyclo-L-prolyl-L-leucine, Gancidin W, L-Leucyl-L-proline lactam, L-Leucyl-L-prolyl lactam, Maculosin 6
IUPAC-namn (3S,8aS)-3-(2-methylpropyl)-2,3,6,7,8,8a-hexahydropyrrolo[1,2-a]pyrazine-1,4-dione
LEDER CC(C)C[C@@H]1NC(=O)[C@@H]2CCCN2C1=O
Specifikationer
Kemiskt namn eller material Cyclo(L-prolyl-L-leucyl)
Kvantitet 50 mg
InChI formel InChI=1S/C11H18N2O2/c1-7(2)6-8-11(15)13-5-3-4-9(13)10(14)12-8/h7-9H,3-6H2,1-2H3,(H,12,14)/t8-,9-/m0/s1
Formel vikt 210.1368