missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4F8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20011-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to CYP4F8. The antibody has no cross reaction to other CYP4F members.
CYP4F8 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and functions as a 19-hydroxylase of prostaglandins in seminal vesicles.Specifications
CYP4F8 | |
Polyclonal | |
Unconjugated | |
CYP4F8 | |
CPF8, CYP4F8, CYPIVF8, Cytochrome P450 4F8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11283 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P98187 | |
CYP4F8 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CYP4F8 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction