missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12365-1-AP-20UL
This item is not returnable.
View return policy
Description
Cytochrome b5 is a membrane-bound member of the cytochrome b family. A heme protein that functions as an electron carrier for many membrane-bound oxygenases, cytochrome b5 possesses two heme groups, which are not covalently attached to the protein. Two isoforms of cytochrome b5, a microsomal membrane-bound form and a cytoplasmic form, are produced by alternative splicing. Mutations in cytochrome b5 are associated with Leber′s hereditary optic neuropathy and with myopathy.Specifications
Cytochrome b5 | |
Polyclonal | |
Unconjugated | |
CYB5A | |
CYB5, CYB5A, Cytochrome b5, MCB5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109672, 1528, 64001 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P00167, P00173, P56395 | |
Cyb5a | |
Cytochrome b5 Fusion Protein Ag3038 | |
20 ÎĽL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytochrome b5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction