Learn More
Cytohesin 2 Mouse anti-Human, Unconjugated, Clone: 3A7A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67185-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839685
  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The Cytohesin 2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cytohesin 2. This antibody recognizes human, mouse, rat antigen. The Cytohesin 2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Cytohesin 2
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
Immunofluorescence, Immunohistochemistry, Western Blot
3A7A1
Unconjugated
150 μL
9266
IgG2a