missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK

Specifications

Antigen Cytokeratin 8
Concentration 0.2 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886733
View Documents
Proteintech
10384-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16876733
View Documents
Proteintech
10384-1-AP-150UL
150 ÎĽL N/A
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
Specifications

Specifications

Cytokeratin 8
Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
P05787, P11679, Q10758
16691, 25626, 3856
Cytokeratin 8 Fusion Protein Ag0488
Primary
-20°C
KRT8
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CARD2, CK 8, CK8, CYK8, Cytokeratin 8, K2C8, K8, keratin 8, KO, KRT8, Type II keratin Kb8
Krt8
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytokeratin 8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.