missing translation for 'onlineSavingsMsg'
Learn More
D(+)-Glucose Monohydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ Product Code.: 10529190

D(+)-Glucose Monohydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. p-7073040 Shop All Fisher Chemical Products
1 kg, Plastic powder jar, 1kg
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 10529190

Brand: Fisher Chemical G/0400/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

D(+)-Glucose Monohydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O
MDL Number 149450

Specifications

Color White
pH 5.9
Packaging Plastic powder jar
Quantity 1 kg
Formula Weight 198.18g/mol
Physical Form Solid
Product Identifier
  • D(+)-Glucose monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D(+)-Glucose Monohydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.