Läs mer
D-Homoanalog of Triamcinolone, TRC
Produktkod. 30136524
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136524 50 mg
30132705 100 mg
30010357 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136524 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH58501050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 130063-07-9
Molekylformel C21H27FO6
Molekylvikt (g/mol) 394.43
Synonym (11beta,16alpha,17alpha)-9-Fluoro-11,16,17-trihydroxy-17-(hydroxymethyl)-D-homoandrosta-1,4-diene-3,17a-dione, (2R,3R,4aS,4bS,10aS,10bR,11S,12aS)-10b-Fluoro-3,4,4a,5,6,10a,10b,11,12,12a-decahydro-2,3,11-trihydroxy-2-(hydroxymethyl)-10a,12a-dimethyl-1,8(2H,4bH)-chrysenedione;
IUPAC-namn (2R,3R,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-2,3,11-trihydroxy-2-(hydroxymethyl)-10a,12a-dimethyl-3,4,4a,4b,5,6,11,12-octahydrochrysene-1,8-dione
LEDER C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@](O)(CO)C2=O
Specifikationer
Kemiskt namn eller material D-Homoanalog of Triamcinolone
Kvantitet 50 mg
InChI formel InChI=1S/C21H27FO6/c1-18-9-16(26)21(22)13(4-3-11-7-12(24)5-6-19(11,21)2)14(18)8-15(25)20(28,10-23)17(18)27/h5-7,13-16,23,25-26,28H,3-4,8-10H2,1-2H3/t13-,14-,15+,16-,18-,19-,20+,21-/m0/s1
Formel vikt 394.1792