missing translation for 'onlineSavingsMsg'
Learn More

DCDC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1706.43 SEK - 4863.32 SEK

Specifications

Antigen DCDC2
Concentration 0.33 mg/mL
Applications Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16885295
View Documents
Proteintech
26978-1-AP-20UL
20 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16875295
View Documents
Proteintech
26978-1-AP-150UL
150 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This gene encodes a member of the doublecortin family. The protein encoded by this gene contains two doublecortin domains. The doublecortin domain has been demonstrated to bind tubulin and enhance microtubule polymerization. Mutations in this gene have been associated with Reading Disability, also referred to as developmental dyslexia. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

DCDC2
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse
Q5DU00, Q9UHG0
195208, 51473
DCDC2 Fusion Protein Ag25577
Primary
-20°C
DCDC2
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DCDC2, DCDC2A, KIAA1154, Protein RU2S, RU2, RU2S
DCDC2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DCDC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.