missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DDB1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888413
|
Proteintech
11380-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878413
|
Proteintech
11380-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to DDB1.
This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.Specifications
DDB1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q16531, Q3U1J4, Q9ESW0 | |
13194, 1642, 64470 | |
DDB1 Fusion Protein Ag1901 | |
Primary | |
-20°C | |
DDB1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor | |
DDB1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DDB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech