Learn More
DDI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13968-1-AP-150UL
 View more versions of this product
Product Code. 16882414
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
Specifications
Specifications
DDI1
Polyclonal
Unconjugated
DDI1
DDI1, Protein DDI1 homolog 1
Rabbit
Antigen Affinity Chromatography
RUO
367012, 414301, 71829
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A0JPP7, Q8WTU0, Q9DAF3
Ddi1
DDI1 Fusion Protein Ag4992
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DDI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated