Learn More
DDI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13968-1-AP-150UL
 View more versions of this product
Product Code. 16882414
  • 4652.90 SEK / 150µL
Description
Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
Specifications
Specifications
DDI1
Polyclonal
Unconjugated
DDI1
DDI1, Protein DDI1 homolog 1
Rabbit
Antigen Affinity Chromatography
RUO
367012, 414301, 71829
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A0JPP7, Q8WTU0, Q9DAF3
Ddi1
DDI1 Fusion Protein Ag4992
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DDI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated