missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDOST Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14916-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a component of the oligosaccharyltransferase complex which catalyzes the transfer of high-mannose oligosaccharides to asparagine residues on nascent polypeptides in the lumen of the rough endoplasmic reticulum. The protein complex co-purifies with ribosomes. The product of this gene is also implicated in the processing of advanced glycation endproducts (AGEs), which form from non-enzymatic reactions between sugars and proteins or lipids and are associated with aging and hyperglycemia.Specifications
DDOST | |
Polyclonal | |
Unconjugated | |
DDOST | |
AGE R1, DDOST, DDOST 48 kDa subunit, KIAA0115, OK/SW cl.45, OST, OST48, WBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13200, 1650, 313648 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P39656, Q641Y0 | |
Ddost | |
DDOST Fusion Protein Ag6727 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DDOST Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction