Learn More
Eurofins DiscoverX DDR2 Protein, Active
Active, N-terminal His6-tagged, recombinant, human DDR2 amino acids 467-end, for use in Kinase Assays.

6046.30 SEK - 46147.75 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 48 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17360374
![]() |
Eurofins DiscoverX
14-579 |
10 ÎĽg |
6046.30 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17370374
![]() |
Eurofins DiscoverX
14-579M |
250 ÎĽg |
46147.75 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Receptor tyrosine kinases (RTKs) play a key role in the communication of cells with their microenvironment. These molecules are involved in the regulation of cell growth, differentiation, and metabolism. In several cases the biochemical mechanism by which RTKs transduce signals across the membrane has been shown to be ligand induced receptor oligomerization and subsequent intracellular phosphorylation. This autophosphorylation leads to phosphorylation of cytosolic targets as well as association with other molecules, which are involved in pleiotropic effects of signal transduction. RTKs have a tripartite structure with extracellular, transmembrane, and cytoplasmic regions. This gene encodes a member of a novel subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain. Alternative splicing in the 5' UTR results in multiple transcript variants encoding the same protein.
GenBank NM_006182. The recombinant protein contains the amino acid substitution S642A (native coordinates) with respect toGenBank NM_006182. This conflict is reported inGenBank accession n° NM_022563, BC052998, BQ678490, BU168475 and BQ682363.
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal 6His | |
Truncated | |
DDR2, active |
48 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥46% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.