missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15769-1-AP-150UL
This item is not returnable.
View return policy
Description
METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.Specifications
DDX24 | |
Polyclonal | |
Unconjugated | |
DDX24 | |
DDX24, DEAD box protein 24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27225, 373065, 57062 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ESV0, Q9GZR7 | |
Ddx24 | |
DDX24 Fusion Protein Ag8407 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DDX24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction