missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DEF6 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888393
|
Proteintech
11369-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878393
|
Proteintech
11369-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.Specifications
DEF6 | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q8C2K1, Q9H4E7 | |
23853, 50619 | |
DEF6 Fusion Protein Ag1925 | |
Primary | |
-20°C | |
DEF6 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DEF 6, DEF6, IBP, IRF4 binding protein, SLAT | |
DEF6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DEF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech