Learn More
DEFA1 Mouse anti-Human, Unconjugated, Clone: 1A2B3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67156-1-IG-20UL
 View more versions of this product
Product Code. 16889625
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The DEFA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DEFA1. This antibody recognizes human antigen. The DEFA1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
DEFA1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1A2B3
Unconjugated
20 μL
1667
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DEFA1 Mouse anti-Human, Unconjugated, Clone: 1A2B3, Proteintech > 20 μL