Learn More
LNI Swissgas Deionized Water Filter for HG BASIC/PRO/KUBE Last Series with Silver
Deionized water filter for HG BASIC/PRO/KUBE last series with silver.
Brand:  LNI Swissgas 6930.00.091
Product Code. 17812329
  • 1295.00 SEK / Each
This item is not returnable. View return policy
Description
Description
  • For hydrogen range
  • Replace every 6 months
Specifications
Specifications
Cartridge
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LNI Swissgas Deionized Water Filter for HG BASIC/PRO/KUBE Last Series with Silver > LNI Swissgas Deionized Water Filter