Läs mer
Delavirdine Mesylate, TRC
Produktkod. 30135531
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135531 10 mg
30007259 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135531 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23063010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147221-93-0
Molekylformel C22 H28 N6 O3 S . C H4 O3 S
Molekylvikt (g/mol) 552.67
Synonym Delavirdine Mesylate, Piperazine, 1-[3-[(1-methylethyl)amino]-2-pyridinyl]-4-[[5-[(methylsulfonyl)amino]-1H-indol-2-yl]carbonyl]-, monomethanesulfonate (9CI), Delavirdine mesylate, Delavirdine methane sulfonate, Rescripta, Rescriptor, U 90152E, U 90152S, U 90152T
IUPAC-namn methanesulfonic acid;N-[2-[4-[3-(propan-2-ylamino)pyridin-2-yl]piperazine-1-carbonyl]-1H-indol-5-yl]methanesulfonamide
LEDER CC(C)Nc1cccnc1N2CCN(CC2)C(=O)c3cc4cc(NS(=O)(=O)C)ccc4[nH]3.CS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Delavirdine mesylate
Kvantitet 10 mg
InChI formel InChI=1S/C22H28N6O3S.CH4O3S/c1-15(2)24-19-5-4-8-23-21(19)27-9-11-28(12-10-27)22(29)20-14-16-13-17(26-32(3,30)31)6-7-18(16)25-20;1-5(2,3)4/h4-8,13-15,24-26H,9-12H2,1-3H3;1H3,(H,2,3,4)
Formel vikt 552.1825