Learn More
Cole-Parmer™ Delrin™ Manual Pinch Valve
675.00 SEK - 790.00 SEK
Specifications
Product Type | Manual Pinch Valve |
---|---|
Material | Delrin |
For Use With (Application) | Ideal for use in pharmaceutical and food industries where eliminating contamination is critical |
Packaging Quantity | 1 Pack |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
11747288
|
Cole-Parmer™
98002-10 |
Fits 5/16 in. to 3/8 in. Tube O.D. |
710.00 SEK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
11776338
|
Cole-Parmer™
98002-00 |
Fits 5/32 in. to 1/4 in. Tube O.D. |
675.00 SEK
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
11757288
|
Cole-Parmer™
98002-20 |
Fits 7/16 in. to 1/2 in. Tube O.D. |
790.00 SEK
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
Specifications
Manual Pinch Valve | |
Delrin | |
Ideal for use in pharmaceutical and food industries where eliminating contamination is critical | |
1 Pack |