Learn More
Thermo Scientific Chemicals Delta Sleep-Inducing Peptide Product Code.: 15435599
Thermo Scientific Chemicals Delta Sleep-Inducing Peptide
Product Code. p-5384677
1 mg, 1mg
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 15435599
Brand: Thermo Scientific Chemicals J63334.M^
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Common Name DSIP
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 848.81
Product Type Delta Sleep-Inducing Peptide
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Product Code
Lot NumberG08X078 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeJ63334.M^,J63334.#0
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Delta Sleep-Inducing Peptide >