missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dermatopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Dermatopontin |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Produktcode | Marke | Quantity | Preis | Menge & VerfĂĽgbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & VerfĂĽgbarkeit | |||||
16883584
|
Proteintech
14777-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873584
|
Proteintech
14777-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.Spezifikation
Dermatopontin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q07507, Q9QZZ6 | |
1805, 289178, 56429 | |
Dermatopontin Fusion Protein Ag6200 | |
Primary | |
-20°C | |
DPT |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Dermatopontin, DPT, TRAMP | |
Dpt | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
Dermatopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech