missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dermatopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10537-1-AP-150UL
This item is not returnable.
View return policy
Description
Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.Specifications
Dermatopontin | |
Polyclonal | |
Unconjugated | |
DPT | |
Dermatopontin, DPT, TRAMP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1805, 289178, 56429 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q07507, Q9QZZ6 | |
Dpt | |
Dermatopontin Fusion Protein Ag0801 | |
150 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dermatopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction