missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[Des-Arg9]-Bradykinin, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
1mg
5mg
Unit Size:
1mg
5mg
Specifications
Specifications
CAS | 15958-92-6 |
Color | White |
Format | Lyophilized Powder |
Molecular Weight (g/mol) | 904.04 |
Product Type | [Des-Arg9]-Bradykinin |
Quantity | 1mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
[Des-Arg9]-Bradykinin, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction