Läs mer
Desmethyl Metsulfuron-methyl, TRC
Produktkod. 17953755
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17953755 50 mg
17903145 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17953755 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29235050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126312-31-0
Molekylformel C13H13N5O6S
Molekylvikt (g/mol) 367.34
Synonym 2-[[[[(3,4-Dihydro-6-methyl-4-oxo-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]benzoic Acid Methyl Ester, 2-[[[[(1,4-Dihydro-6-methyl-4-oxo-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]benzoic Acid Methyl Ester;
LEDER COC(=O)c1ccccc1S(=O)(=O)NC(=O)NC2=NC(=NC(=O)N2)C
Specifikationer
Kemiskt namn eller material Desmethyl Metsulfuron-methyl
Kvantitet 50 mg
InChI formel InChI=1S/C13H13N5O6S/c1-7-14-11(16-12(20)15-7)17-13(21)18-25(22,23)9-6-4-3-5-8(9)10(19)24-2/h3-6H,1-2H3,(H3,14,15,16,17,18,20,21)