missing translation for 'onlineSavingsMsg'
Learn More

Dextrin, Commercial grade Type II, MP Biomedicals™

Catalog Number 11435532
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 9004-53-9
Color White to Yellow
Molecular Formula C18H32O16
Quantity 500 g
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 504.438
PubChem CID 62698
Grade Commercial Type II
Physical Form Powder
Chemical Name or Material Dextrin
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'