missing translation for 'onlineSavingsMsg'
Learn More
Dextrinized Corn Starch, MP Biomedicals Product Code.: 11451765

Dextrinized Corn Starch, MP Biomedicals

Product Code. p-7112298 Shop All MP Biomedical Products
5 lb., 5lb
Click to view available options
Quantity:
5 lb.
25 lb.
Unit Size:
25lb
5lb
This item is not returnable. View return policy

Product Code. 11451765

Brand: MP Biomedicals 0296042905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • A component in animal diet

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Maltodextin

Specifications

Color White
Quantity 5 lb.
Physical Form Powder
Chemical Name or Material DEXTRINIZED CORN STARCH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dextrinized Corn Starch, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.