missing translation for 'onlineSavingsMsg'
Learn More
Dextrorotation nimorazole phosphate ester, MedChemExpress Product Code.: 16493617

Dextrorotation nimorazole phosphate ester, MedChemExpress

Product Code. 16493617
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493617

Brand: Medchem Express HY18716

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dextrorotation nimorazole phosphate ester is an anti-anaerobic and anti-parasitic agent.

Dextrorotation nimorazole phosphate ester is an anti-anaerobic and anti-parasitic agent.

Specifications

Chemical Name or Material Dextrorotation nimorazole phosphate ester
Physical Form Solid
CAS 1124347-33-6
Color Red
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H19N4O7P
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 32 mg/mL (91.36 mM)
SMILES CC1=NC=C([N+]([O-])=O)N1C[C@H](OP(O)(O)=O)CN2CCOCC2
Molecular Weight (g/mol) 350.26
Formula Weight 350.26
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Dextrorotation nimorazole phosphate ester, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.