missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11561-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.Specifications
DGAT1 | |
Polyclonal | |
Unconjugated | |
DGAT1 | |
ACAT related gene product 1, AGRP1, ARGP1, DGAT, DGAT1, Diglyceride acyltransferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13350, 84497, 8694 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75907, Q9ERM3, Q9Z2A7 | |
Dgat1 | |
DGAT1 Fusion Protein Ag2123 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |