missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4863.32 SEK
Specifications
Antigen | DGAT1 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.Specifications
DGAT1 | |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O75907, Q9ERM3, Q9Z2A7 | |
13350, 84497, 8694 | |
DGAT1 Fusion Protein Ag2123 | |
Primary | |
-20°C | |
DGAT1 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACAT related gene product 1, AGRP1, ARGP1, DGAT, DGAT1, Diglyceride acyltransferase | |
Dgat1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DGAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech