missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13900-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction.Specifications
DHDH | |
Polyclonal | |
Unconjugated | |
DHDH | |
2DD, D xylose 1 dehydrogenase, D xylose NADP dehydrogenase, DHDH, HUM2DD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27294, 691002, 71755 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9DBB8, Q9UQ10 | |
DHDH | |
DHDH Fusion Protein Ag4866 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DHDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction