Learn More
DHRS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20172-1-AP-150UL
 View more versions of this product
Product Code. 16889674
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
DHRS11 gene ontology annotations related to this gene include oxidoreductase activity.
Specifications
Specifications
DHRS11
Polyclonal
Unconjugated
DHRS11
ARPG836, DHRS11, SDR24C1
Rabbit
Antigen Affinity Chromatography
RUO
192970, 360583, 79154
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3U0B3, Q6UWP2
Dhrs11
DHRS11 Fusion Protein Ag14005
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DHRS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated